Learn More
Biomatik Corporation Human Fibulin-7 (Fbln7) Elisa Kit
Supplier:  Biomatik Corporation EKC33721
Encompass_Preferred
Human Fibulin-7 (Fbln7) Elisa Kit for detecting Fibulin 7 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q53RD9; FBLN7. Target Synonyms: FBLN7; FBLN7_HUMAN; FIBL-7; Fibl7; Fibulin-7; TM14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3968