Learn More
Biomatik Corporation Human Filamin-A (Flna) Elisa Kit
Supplier:  Biomatik Corporation EKC33727
Encompass_Preferred
Human Filamin-A (Flna) Elisa Kit for detecting Filamin A, Alpha (Actin Binding Protein 280) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 18.75-1200pg/ml. Sensitivity: 4.68pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P21333; FLNA. Target Synonyms: ABP 280; ABP-280; Actin-binding protein 280; Alpha filamin; Alpha-filamin; APBX; CSBS; CVD1; Endothelial actin binding protein; Endothelial actin-binding protein; Filamin 1; Filamin A alpha; Filamin A; Filamin-1; Filamin-A; FLN; FLN-A; FLN1; FLNA; FLNA_HUMAN; FMD; MNS; NHBP; Non muscle filamin; Non-muscle filamin; OPD; OPD1; OPD2; XLVD; XMVD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3971