Learn More
Biomatik Corporation Human G protein-coupled bile acid receptor 1(GPBAR1) ELISA Kit
Supplier:  Biomatik Corporation EKC33767
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q8TDU6. Target Synonyms: BG37, GPCR, GPCR19, GPR131, M-BAR, MGC40597, TGR5, G-protein coupled bile acid receptor BG37|membrane bile acid receptor|membrane-type receptor for bile acids. Restriction: For Research Use Only.
Catalog No. 50-229-3981