Learn More
Biomatik Corporation Human Glucosylceramidase (Gba) Elisa Kit
Supplier:  Biomatik Corporation EKC33834
Encompass_Preferred
Human Glucosylceramidase (Gba) Elisa Kit for detecting Glucosidase, Beta; Acid (Includes Glucosylceramidase) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04062; GBA. Target Synonyms: Acid beta glucosidase; Acid beta-glucosidase; Alglucerase; Beta glucocerebrosidase; BETA GLUCOSIDASE; ACID; Beta-glucocerebrosidase; betaGC; D glucosyl N acylsphingosine glucohydrolase; D-glucosyl-N-acylsphingosine glucohydrolase; EC 3.2.1.45; GBA; Gba protein; GBA1; GC; GCase; GCB; GLCM_HUMAN; GLUC; Glucocerebrosidase (alt.); Glucocerebrosidase; GLUCOCEREBROSIDASE PSEUDOGENE; Glucosidase beta; Glucosidase; beta; acid; Glucosidase; beta; acid (includes glucosylceramidase); Glucosylceramidase; Imiglucerase; Lysosomal glucocerebrosidase; OTTHUMP00000033992. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3993