Learn More
Biomatik Corporation Human Glutamic Acid Decarboxylase, Gad Elisa Kit
Supplier:  Biomatik Corporation EKC33847
Encompass_Preferred
Human Glutamic Acid Decarboxylase, Gad Elisa Kit for detecting Glutamate Decarboxylase 1 (Brain, 67Kda) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q99259; GAD1. Target Synonyms: 67 kDa glutamic acid decarboxylase; CPSQ1; DCE1; DCE1_HUMAN; EC 4.1.1.15; FLJ45882; GAD 1; GAD 67; GAD; GAD-67; GAD1; Glutamate decarboxylase 1 (brain; 67kDa); Glutamate decarboxylase 1; Glutamate decarboxylase 1 brain 67kD; Glutamate decarboxylase 1 brain 67kDa; Glutamate decarboxylase 67 kDa isoform; Glutamate decarboxylase; brain; 67-KD; OTTHUMP00000041054; OTTHUMP00000041055; OTTHUMP00000228572; SCP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3995