Learn More
Biomatik Corporation Human Glypican 5 (Gpc5) Elisa Kit
Supplier:  Biomatik Corporation EKC33880
Encompass_Preferred
Human Glypican 5 (Gpc5) Elisa Kit for detecting Glypican 5 in serum, plasma, ascitic fluid. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P78333; GPC5. Target Synonyms: bA93M14.1; glypican 5; Glypican 5 precursor; glypican proteoglycan 5; Glypican-5; GPC5; GPC5_HUMAN; Secreted glypican-5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4005