Learn More
Biomatik Corporation Human Gremlin-2 (Grem2) Elisa Kit
Supplier:  Biomatik Corporation EKC33906
Encompass_Preferred
Human Gremlin-2 (Grem2) Elisa Kit for detecting Gremlin 2, Cysteine Knot Superfamily, Homolog (Xenopus Laevis) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-2500pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q9H772; GREM2. Target Synonyms: BMP antagonist 2; CKTSF1B2; Cysteine knot superfamily 1; Cysteine knot superfamily 1 BMP antagonist 2; DAN domain family member 3; DAND 3; DAND3; GREM 2; Grem2; GREM2_HUMAN; Gremlin 2; Gremlin 2 cysteine knot superfamily homolog; Gremlin-2; Gremlin2; PRDC; Protein related to DAN and cerberus. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4012