Learn More
Biomatik Corporation Human Gtpase Kras (Kras) Elisa Kit
Supplier:  Biomatik Corporation EKC33926
Encompass_Preferred
Human Gtpase Kras (Kras) Elisa Kit for detecting V-Ki-Ras2 Kirsten Rat Sarcoma Viral Oncogene Homolog in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01116; KRAS. Target Synonyms: c Ki ras2; c Kirsten ras protein; c-K-ras; c-Ki-ras; Cellular c Ki ras2 proto oncogene; Cellular transforming proto oncogene; CFC2; cK Ras; GTPase KRas; K RAS p21 protein; K RAS2A; K RAS2B; K RAS4A; K RAS4B; K-Ras 2; KI RAS; Ki-Ras; KIRSTEN MURINE SARCOMA VIRUS 2; Kirsten rat sarcoma 2 viral (v Ki ras2) oncogene homolog; Kirsten rat sarcoma viral oncogene homolog; KRAS; KRAS proto oncogene; GTPase; KRAS1; KRAS2; N-terminally processed; NS; NS3; Oncogene KRAS2; p21ras; PR310 c K ras oncogene; PR310 cK ras oncogene; RALD; RASK_HUMAN; RASK2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4017