Learn More
Biomatik Corporation Human Heme Oxygenase 1, Ho-1 Elisa Kit
Supplier:  Biomatik Corporation EKC33968
Encompass_Preferred
Human Heme Oxygenase 1, Ho-1 Elisa Kit for detecting Heme Oxygenase (Decycling) 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000ng/ml. Sensitivity: 3.9ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P09601; HMOX1. Target Synonyms: 32 kD; bK286B10; D8Wsu38e; heat shock protein 32 kD; heat shock protein 32kD; Heat shock protein; Heme oxygenase (decycling) 1; Heme oxygenase 1; Hemox; HMOX 1; Hmox; Hmox1; HMOX1_HUMAN; HO 1; HO; HO-1; HO1; Hsp32. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4024