Learn More
Biomatik Corporation Human High Affinity Immunoglobulin Gamma Fc Receptor I (Fcgr1A) Elisa Kit
Supplier:  Biomatik Corporation EKC34023
Encompass_Preferred
Human High Affinity Immunoglobulin Gamma Fc Receptor I (Fcgr1A) Elisa Kit for detecting Fc Fragment Of Igg, High Affinity Ia, Receptor (Cd64) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.2ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P12314; FCGR1A. Target Synonyms: FCGR1A; FCG1; FCGR1; IGFR1; High affinity immunoglobulin gamma Fc receptor I; IgG Fc receptor I; Fc-gamma RI; FcRI; Fc-gamma RIA; FcgammaRIa; CD antigen CD64. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4028