Learn More
Biomatik Corporation Human Histidine-Rich Glycoprotein (Hrg) Elisa Kit
Supplier:  Biomatik Corporation EKC34037
Encompass_Preferred
Human Histidine-Rich Glycoprotein (Hrg) Elisa Kit for detecting Histidine-Rich Glycoprotein in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400ug/ml. Sensitivity: 1.56ug/mL. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04196; HRG. Target Synonyms: HRG; Histidine-rich glycoprotein; Histidine-proline-rich glycoprotein; HPRG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4031