Learn More
Biomatik Corporation Human Insulin-Like 3 (Insl3) Elisa Kit
Supplier:  Biomatik Corporation EKC34144
Encompass_Preferred
Human Insulin-Like 3 (Insl3) Elisa Kit for detecting Insulin-Like 3 (Leydig Cell) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P51460; INSL3. Target Synonyms: INSL 3; INSL3; INSL3_HUMAN; Insulin like 3; insulin like peptide; Leydig cell-specific; insulin-like 3 (Leydig cell); Insulin-like 3 A chain; Ley IL; Ley-I-L; leydig insulin -like hormone; Leydig insulin like peptide; Leydig insulin-like peptide; prepro-INSL3; Relaxin like factor; Relaxin-like factor; relaxin-like factor b; RLF; RLNL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4053