Learn More
Biomatik Corporation Human Integrin Alpha-2 (Itga2) Elisa Kit
Supplier:  Biomatik Corporation EKC34164
Encompass_Preferred
Human Integrin Alpha-2 (Itga2) Elisa Kit for detecting Integrin, Alpha 2 (Cd49B, Alpha 2 Subunit Of Vla-2 Receptor) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.235-15ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P17301; ITGA2. Target Synonyms: BR; CD 49b; CD49 antigen like family member B; CD49 antigen-like family member B; CD49b; CD49b antigen; Collagen receptor; DX5; Glycoprotein Ia deficiency included; GP Ia; GP Ia deficiency; included; GPIa; HPA 5 included; HPA5 included; Human platelet alloantigen system 5; Integrin alpha 2; Integrin alpha II; Integrin alpha-2; Integrin; alpha 2 (CD49B alpha 2 subunit of VLA 2 receptor); ITA2_HUMAN; ITGA2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4055