Learn More
Biomatik Corporation Rat VGF Nerve Growth Factor Inducible (VGF) ELISA Kit
Supplier:  Biomatik Corporation EKU08141
Encompass_Preferred
Rat VGF Nerve Growth Factor Inducible (VGF) ELISA Kit for detecting VGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 46.88-3,000pg/ml. Sensitivity: 18.34pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20156. Gene ID: 29461. Target Synonyms: NERP1; NERP2; Neuro-Endocrine Specific Protein VGF; Neuroendocrine Regulatory Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3002