Learn More
Biomatik Corporation Human V-Rel Reticuloendotheliosis Viral Oncogene Homolog B (RELB) ELISA Kit
Supplier:  Biomatik Corporation EKU08194
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.063ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q01201. Target Synonyms: REL-B; I-REL; Transcription Factor RelB; Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells 3. Restriction: For Research Use Only.
Catalog No. 50-229-3008