Learn More
Biomatik Corporation Human Wingless Type MMTV Integration Site Family, Member 5B (WNT5B) ELISA Kit
Supplier:  Biomatik Corporation EKU08217
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 1.56-100ng/mL. Sensitivity: 0.55ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9H1J7. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-3010