Learn More
Biomatik Corporation Mouse WNT1 Inducible Signaling Pathway Protein 1 (WISP1) ELISA Kit
Supplier:  Biomatik Corporation EKU08225
Encompass_Preferred
Mouse WNT1 Inducible Signaling Pathway Protein 1 (WISP1) ELISA Kit for detecting WISP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O54775. Gene ID: 22402. Target Synonyms: CCN4; WISP1c; WISP1i; WISP1tc; CCN family member 4; Wnt-1-induced secreted protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3012