Learn More
Biomatik Corporation Mouse Xanthine Dehydrogenase (XDH) ELISA Kit
Supplier:  Biomatik Corporation EKU08233
Encompass_Preferred
Mouse Xanthine Dehydrogenase (XDH) ELISA Kit for detecting XDH in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q00519. Gene ID: 22436. Target Synonyms: XO; XOR; XOD; Xanthine Oxidase; NAD+-Xanthine Dehydrogenase; Xanthine-NAD+ Oxidoreductase; Xanthine/NAD+ Oxidoreductase; Xanthine Oxidoreductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3014