Learn More
Biomatik Corporation Human YY1 Transcription Factor (YY1) ELISA Kit
Supplier:  Biomatik Corporation EKU08249
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.060ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P25490. Target Synonyms: NF-E1; DELTA; UCRBP; YIN-YANG-1; INO80S; INO80 Complex Subunit S; Yin and Yang 1 Protein; Delta transcription factor; INO80 complex subunit S. Restriction: For Research Use Only.
Catalog No. 50-229-3016