Learn More
Biomatik Corporation Human Tumor Necrosis Factor Receptor Superfamily, Member 7 (TNFRSF7) ELISA Kit
Supplier:  Biomatik Corporation EKU08307
Encompass_Preferred
Human Tumor Necrosis Factor Receptor Superfamily, Member 7 (TNFRSF7) ELISA Kit for detecting TNFRSF7 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P26842. Gene ID: 939. Target Synonyms: CD27; S152; T14; Tp55; CD27L receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3019