Learn More
Biomatik Corporation Rat Phospholipase A2, Calcium Independent (iPLA2) ELISA Kit
Supplier:  Biomatik Corporation EKU08314
Encompass_Preferred
Rat Phospholipase A2, Calcium Independent (iPLA2) ELISA Kit for detecting iPLA2 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97570. Gene ID: 360426. Target Synonyms: PLA2G6; CaI-PLA2; GVI; INAD1; PNPLA9; Ca2+ Independent PLA2; 85 kDa Calcium-Independent Phospholipase A2; Phospholipase A2,group VI (Cytosolic,Calcium-Independent). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3021