Learn More
Biomatik Corporation Mouse Pyruvate Dehydrogenase Kinase Isozyme 2 (PDK2) ELISA Kit
Supplier:  Biomatik Corporation EKU08340
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9JK42. Target Synonyms: PDHK2; [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 2, mitochondrial. Restriction: For Research Use Only.
Catalog No. 50-229-3027