Learn More
Biomatik Corporation Mouse Integrin Beta 6 (ITGb6) ELISA Kit
Supplier:  Biomatik Corporation EKU08450
Encompass_Preferred
Mouse Integrin Beta 6 (ITGb6) ELISA Kit for detecting ITGb6 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z0T9. Gene ID: 16420. Target Synonyms: ITG-B6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3049