Learn More
Biomatik Corporation Mouse Collagen Type III Alpha 1 (COL3a1) ELISA Kit
Supplier:  Biomatik Corporation EKU08476
Encompass_Preferred
Mouse Collagen Type III Alpha 1 (COL3a1) ELISA Kit for detecting COL3a1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 4.7-300ng/ml. Sensitivity: 2.3ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11087. Gene ID: 12842. Target Synonyms: COL3-A1; COL3A1; EDS4A; Collagen Type III Alpha 1; Ehlers-Danlos Syndrome Type IV Autosomal Dominant; Collagen Alpha-1 (III) chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3051