Learn More
Biomatik Corporation Rat Myoglobin (MYO) CLIA Kit
Supplier:  Biomatik Corporation EKU08873
Encompass_Preferred
Rat Myoglobin (MYO) CLIA Kit for detecting MYO in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 54.9-40,000pg/ml. Sensitivity: 19.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9QZ76. Gene ID: 59108. Target Synonyms: PVALB; MB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3070