Learn More
Biomatik Corporation Human Alpha-1-Antitrypsin (a1AT) CLIA Kit
Supplier:  Biomatik Corporation EKU08944
Encompass_Preferred
Human Alpha-1-Antitrypsin (a1AT) CLIA Kit for detecting a1AT in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.55-400ng/ml. Sensitivity: 0.22ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P01009. Gene ID: 5265. Target Synonyms: SERPINA1; SPAAT; A1-AT; PI; Serpin Peptidase Inhibitor,Clade A (Alpha-1 Antiproteinase/AntiTrypsin) Member 1; Alpha-1 protease inhibitor; Short peptide from AAT; Serpin A1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3075