Learn More
Biomatik Corporation Human Chemokine C-X-C-Motif Receptor 6 (CXCR6) ELISA Kit
Supplier:  Biomatik Corporation EKU09036
Encompass_Preferred
Human Chemokine C-X-C-Motif Receptor 6 (CXCR6) ELISA Kit for detecting CXCR6 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O00574. Gene ID: 10663. Target Synonyms: CD186; BONZO; STRL33; TYMSTR; G-protein coupled receptor STRL33; G-protein coupled receptor bonzo. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3083