Learn More
Biomatik Corporation Rat Permeability Glycoprotein (Pgp) ELISA Kit
Supplier:  Biomatik Corporation EKU09074
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.056ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P43245. Target Synonyms: CD243; ABCB1; ABC20; CLCS; GP170; MDR1; PGY1; P-Glycoprotein; Multidrug resistance protein 1; ATP-Binding Cassette,Sub-Family B(MDR/TAP),Member 1; Colchicin Sensitivity. Restriction: For Research Use Only.
Catalog No. 50-229-3089