Learn More
Biomatik Corporation Human/Mouse/Rat Tubulin Beta (TUBb) ELISA Kit
Supplier:  Biomatik Corporation EKU09094
Encompass_Preferred
Human/Mouse/Rat Tubulin Beta (TUBb) ELISA Kit for detecting TUBb in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07437. Gene ID: 203068. Target Synonyms: TUB-B; TUBB5; TUBB1; M40. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3092