Learn More
Biomatik Corporation Human Crystallin Lambda 1 (CRYl1) ELISA Kit
Supplier:  Biomatik Corporation EKU09216
Encompass_Preferred
Human Crystallin Lambda 1 (CRYl1) ELISA Kit for detecting CRYl1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y2S2. Gene ID: 51084. Target Synonyms: GDH; Lambda-CRY; Gul3DH; L-Gulonate 3-Dehydrogenase; Lambda-Crystallin Homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3101