Learn More
Biomatik Corporation Human Tricellulin (TRIC) ELISA Kit
Supplier:  Biomatik Corporation EKU09314
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.116ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: MARVELD2; DFNB49; MARVD2; MRVLDC2; MARVEL Domain Containing Protein 2; Deafness,Autosomal Recessive 49. Restriction: For Research Use Only.
Catalog No. 50-229-3115