Learn More
Biomatik Corporation Rat Substance P (SP) CLIA Kit
Supplier:  Biomatik Corporation EKU09455
Encompass_Preferred
Rat Substance P (SP) CLIA Kit for detecting SP in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.9-1,000pg/ml. Sensitivity: 1.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P06767. Gene ID: 24806. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3129