Learn More
Biomatik Corporation Human Anti-Mullerian Hormone (AMH) CLIA Kit
Supplier:  Biomatik Corporation EKU09492
Encompass_Preferred
Human Anti-Mullerian Hormone (AMH) CLIA Kit for detecting AMH in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 195.3-50,000pg/ml. Sensitivity: 74.3pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P03971. Gene ID: 268. Target Synonyms: MIF; MIH; MIS; Müllerian Inhibiting Factor; Müllerian Inhibiting Hormone; Müllerian Inhibiting Substance. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3134