Learn More
Biomatik Corporation Mouse Interleukin 17C (IL17C) CLIA Kit
Supplier:  Biomatik Corporation EKU09552
Encompass_Preferred
Mouse Interleukin 17C (IL17C) CLIA Kit for detecting IL17C in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.51pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q8K4C5. Gene ID: 234836. Target Synonyms: Cytokine CX2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3136