Learn More
Biomatik Corporation Human S100 Calcium Binding Protein B (S100B) CLIA Kit
Supplier:  Biomatik Corporation EKU09890
Encompass_Preferred
Human S100 Calcium Binding Protein B (S100B) CLIA Kit for detecting S100B in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P04271. Gene ID: 6285. Target Synonyms: S100-B; NEF; S100Beta; S-100 protein subunit beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3148