Learn More
Biomatik Corporation Human Activating Transcription Factor 1 (ATF1) ELISA Kit
Supplier:  Biomatik Corporation EKU10056
Encompass_Preferred
Human Activating Transcription Factor 1 (ATF1) ELISA Kit for detecting ATF1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P18846. Gene ID: 466. Target Synonyms: EWS-ATF1; FUS/ATF-1; TREB36; Cyclic AMP-Dependent Transcription Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3150