Learn More
Biomatik Corporation Human Synuclein Alpha (SNCa) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU10065
Encompass_Preferred
Human Synuclein Alpha (SNCa) Anti-antigen ELISA Kit for detecting SNCa in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.14ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P37840. Gene ID: 6622. Target Synonyms: SNC-A; aSYN; PD1; PARK1; PARK4; NACP; Non-A Beta Component Of Alzheimer's Disease Amyloid Precursor Protein; Non-A4 component of amyloid precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3153