Learn More
Biomatik Corporation Rat Serum Amyloid A (SAA) ELISA Kit
Supplier:  Biomatik Corporation EKU10219
Encompass_Preferred
Rat Serum Amyloid A (SAA) ELISA Kit for detecting SAA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 39-10,000ng/ml. Sensitivity: 17ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q5M878. Gene ID: 365245. Target Synonyms: SA-A; SAA1; PIG4; TP53I4; Amyloid protein A; Amyloid fibril protein AA; Serum amyloid A-1 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3166