Learn More
Biomatik Corporation Human Synaptosomal Associated Protein 25kDa (SNAP25) ELISA Kit
Supplier:  Biomatik Corporation EKU10236
Encompass_Preferred
Human Synaptosomal Associated Protein 25kDa (SNAP25) ELISA Kit for detecting SNAP25 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P60880. Gene ID: 6616. Target Synonyms: RIC4; SEC9; SNAP; SUP; Super protein; Resistance To Inhibitors Of Cholinesterase 4 Homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3168