Learn More
Biomatik Corporation Mouse Hepcidin (Hepc) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10284
Encompass_Preferred
Mouse Hepcidin (Hepc) High-sensitive ELISA Kit for detecting Hepc in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9EQ21. Gene ID: 84506. Target Synonyms: HAMP; HFE2B; PLTR; LEAP1; Hepcidin Antimicrobial Peptide; Liver-expressed antimicrobial peptide 1; Putative liver tumor regressor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3177