Learn More
Biomatik Corporation Mouse Klotho (KL) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10558
Encompass_Preferred
Mouse Klotho (KL) Wide-range ELISA Kit for detecting KL in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.5-800pg/ml. Sensitivity: 5.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35082. Gene ID: 16591. Target Synonyms: KLa; Klotho Alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3188