Learn More
Biomatik Corporation Human Peroxiredoxin 2 (PRDX2) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10569
Encompass_Preferred
Human Peroxiredoxin 2 (PRDX2) Wide-range ELISA Kit for detecting PRDX2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P32119. Gene ID: 7001. Target Synonyms: PRP; NKEFB; PRXII; TDPX1; TSA; Thioredoxin-Dependent Peroxide Reductase 1; Thiol-Specific Antioxidant 1; Natural Killer-Enhancing Factor B; Thioredoxin Peroxidase 1; Torin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3190