Learn More
Biomatik Corporation Rat Aquaporin 3, Gill Blood Group (AQP3) ELISA Kit
Supplier:  Biomatik Corporation EKU10660
Encompass_Preferred
Rat Aquaporin 3, Gill Blood Group (AQP3) ELISA Kit for detecting AQP3 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P34080. Gene ID: 25386. Target Synonyms: GIL; Aquaglyceroporin-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3202