Learn More
Biomatik Corporation Mouse Complement C3 Convertase (C3 Convertase) ELISA Kit
Supplier:  Biomatik Corporation EKU10741
Encompass_Preferred
Mouse Complement C3 Convertase (C3 Convertase) ELISA Kit for detecting C3 Convertase in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.137ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01027. Gene ID: 12266. Target Synonyms: C4b2a; C3bBb, complement C.hivin.4.hivin2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3213