Learn More
Biomatik Corporation Pig Cytochrome C (CYCS) ELISA Kit
Supplier:  Biomatik Corporation EKU10773
Encompass_Preferred
Pig Cytochrome C (CYCS) ELISA Kit for detecting CYCS in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P62895. Gene ID: 100170131. Target Synonyms: Cyt-C; HCS; CYC; CytC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3218