Learn More
Biomatik Corporation Mouse Dopamine Receptor D2 (DRD2) ELISA Kit
Supplier:  Biomatik Corporation EKU10800
Encompass_Preferred
Mouse Dopamine Receptor D2 (DRD2) ELISA Kit for detecting DRD2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P61168. Gene ID: 13489. Target Synonyms: DR-D2; D2DR; D2R. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3222