Learn More
Biomatik Corporation Human Fibrinogen (FG) ELISA Kit
Supplier:  Biomatik Corporation EKU10820
Encompass_Preferred
Human Fibrinogen (FG) ELISA Kit for detecting FG in plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 370.4-30,000ng/ml. Sensitivity: 145.2ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P02675. Gene ID: 2244. Target Synonyms: F1; FI; Fbg; Coagulation Factor I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3224