Learn More
Biomatik Corporation Rat Free Fatty Acid Receptor 2 (FFAR2) ELISA Kit
Supplier:  Biomatik Corporation EKU10847
Encompass_Preferred
Rat Free Fatty Acid Receptor 2 (FFAR2) ELISA Kit for detecting FFAR2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q76EI6. Gene ID: 292794. Target Synonyms: FFA2R; GPR43; FFA2; GPCR43; G-protein coupled receptor 43. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3231