Learn More
Biomatik Corporation Heat Stable Enterotoxin a (STa) ELISA Kit
Supplier:  Biomatik Corporation EKU10894
Encompass_Preferred
Heat Stable Enterotoxin a (STa) ELISA Kit for detecting STa in biological agents. Species Reactivity: Universal. Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3235