Learn More
Biomatik Corporation Bovine Tumor Necrosis Factor Alpha (TNFa) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10969
Encompass_Preferred
Bovine Tumor Necrosis Factor Alpha (TNFa) High-sensitive ELISA Kit for detecting TNFa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 7.8-500pg/ml. Sensitivity: 2.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q06599. Gene ID: 280943. Target Synonyms: DIF; TNF-A; TNFSF2; Cachectin; Tumor Necrosis Factor Ligand Superfamily Member 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3243